missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389400025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
0.01 max. (water, 1cm, 254nm) | |
5 to 8 | |
Colorless | |
6μS/cm Max. | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Analysis | |
0.89 mPa.s (25°C) | |
2.5 L | |
1 |
Water | |
7732-18-5 | |
100.0°C | |
H2O | |
MFCD00011332 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Plastic Bottle | |
1.0000g/mL | |
1.3334 | |
Liquid |
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Safety and Handling
RUO – Research Use Only