missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, for analysis, Thermo Scientific Chemicals

$49.31 - $244.92

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC389400010
SDS View Documents
Thermo Scientific Chemicals
389400010
1 L Plastic Bottle
Each for $49.31
 
AC389400025
SDS View Documents
Thermo Scientific Chemicals
389400025
2.5 L Plastic Bottle
Each for $85.96
 
AC389400100
SDS View Documents
Thermo Scientific Chemicals
389400100
10 L Plastic Drum
Each for $244.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O

Specifications

0.01 max. (water, 1cm, 254nm)
7732-18-5
100.0°C
H2O
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Plastic Bottle
1.0000g/mL
1.3334
Liquid
5 to 8
Colorless
6μS/cm Max.
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Analysis
0.89 mPa.s (25°C)
1 L
1
Water
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only