missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 389400100

 View more versions of this product

Catalog No. AC389400100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Specifications

0.01 max. (water, 1cm, 254nm)
5 to 8
Colorless
6μS/cm Max.
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Analysis
0.89 mPa.s (25°C)
10 L
1
Water
7732-18-5
100.0°C
H2O
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Plastic Drum
1.0000g/mL
1.3334
Liquid

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only