missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, Honeywell Riedel-de Haën™

CHROMASOLV™, GC-Headspace tested

Supplier:  Honeywell Inc 534636X1L

 View more versions of this product

Catalog No. 60-090-632


Explore available promotions

Description

Description

Honeywell Chromasolv™ GC-headspace solvents:

  • developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • very pure solvents with extremely low concentrations of the defined residual solvents
  • state-of-the art production and application tests

 

Specifications

Specifications

Water
H2O
NONH for all modes of transport
water, dihydrogen oxide, distilled water, purified water, water vapor
O
18.02
CHEBI:15377
Glass Bottle
6 x 1 L
7732-18-5
MFCD00011332
2050024
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
962
18.02g/mol
1.000g/mL at 3.98°C

Chemical Identifiers

7732-18-5
MFCD00011332
962
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
CHEBI:15377
Videos
Safety and Handling

Safety and Handling

RTECSNumber : 231-791-2

SDS
Documents

Documents

Promotions

Promotions