missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, Honeywell Riedel-de Haën™
CHROMASOLV™, GC-Headspace tested
$1,724.05 - $1,724.05
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
60-090-632
|
Honeywell Inc
534636X1L |
6 x 1 L | Glass Bottle |
N/A
|
|
||||
Description
Honeywell Chromasolv™ GC-headspace solvents:
- developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
- very pure solvents with extremely low concentrations of the defined residual solvents
- state-of-the art production and application tests
Chemical Identifiers
7732-18-5 | |
MFCD00011332 | |
962 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
CHEBI:15377 |
Specifications
7732-18-5 | |
MFCD00011332 | |
2050024 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
962 | |
18.02g/mol | |
1.000g/mL at 3.98°C | |
Water |
H2O | |
NONH for all modes of transport | |
water, dihydrogen oxide, distilled water, purified water, water vapor | |
O | |
18.02 | |
CHEBI:15377 | |
Glass Bottle | |
6 x 1 L |
Safety and Handling
RTECSNumber : 231-791-2