missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.015 |
| MDL Number | MFCD00011332 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| PubChem CID | 962 |
| ChEBI | CHEBI:15377 |
| IUPAC Name | oxidane |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
W5SK-1
|
Fisher Chemical
W5SK1 |
1 L | Safe-Cote™ Amber Glass |
Each for $303.67
Case of 6 Each for $1,822.02
|
|
||||
|
W5-1
|
Thermo Fisher Scientific
W51 |
1 L | Wide Neck Amber Bottle |
Each for $257.75
Case of 6 Each for $1,546.50
|
|
||||
|
W5-4
|
Fisher Chemical
BPW54 |
4 L | Glass Bottle |
Each for $318.33
Case of 4 Each for $1,273.32
|
|
||||
|
W5SK-4
|
Thermo Fisher Scientific
FLW5SK4 |
4 L | Safe-Cote™ Amber Glass |
Each for $422.42
Case of 4 Each for $1,689.68
|
|
||||
|
W5N119
|
Thermo Fisher Scientific
W5N119 |
19 L | NOWPak™ I |
Each for $5,763.67
|
|
||||
Description
Improved filtration! HPLC mobile phase for analytical and preparative separations.Chemical Identifiers
| 7732-18-5 | |
| 18.015 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| 962 | |
| oxidane |
| H2O | |
| MFCD00011332 | |
| dihydrogen oxide, dihydrogen monoxide | |
| CHEBI:15377 | |
| O |
Specifications
| 7 | |
| Colorless | |
| 100°C | |
| H2O | |
| dihydrogen oxide, dihydrogen monoxide | |
| O | |
| 18.015 | |
| CHEBI:15377 | |
| Mobile phase for HPLC applications | |
| 1ppm max. | |
| 0.2-micron filtered. | |
| 1 L | |
| Water |
| 7732-18-5 | |
| 0°C | |
| Improved filtration! HPLC mobile phase for analytical and preparative separations. Now filtered through a 0.2 μm filter to remove particulates. | |
| MFCD00011332 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| oxidane | |
| 962 | |
| 18.02 | |
| Safe-Cote™ Amber Glass | |
| 17.5mmHg at 20°C | |
| 1000g/cm³ | |
| Liquid |
Safety and Handling
Low hazard for usual industrial or commercial handling.
Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .
NFPA
Health:0
Flammability:0
Instability:0