missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water (HPLC), Fisher Chemical™

Supplier:  Thermo Fisher Scientific W51

 View more versions of this product

Catalog No. W5-1

Explore available promotions

Description

Description

Improved filtration! HPLC mobile phase for analytical and preparative separations.
Specifications

Specifications

Water
7732-18-5
0°C
Mobile phase for HPLC applications
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
HPLC
1ppm max.
0.2-micron filtered.
1 L
7
Colorless
100°C
Improved filtration! HPLC mobile phase for analytical and preparative separations. Now filtered through a 0.2 μm filter to remove particulates.
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Wide Neck Amber Bottle
17.5mmHg at 20°C
1000g/cm³
Liquid

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
Videos
Safety and Handling

Safety and Handling

Low hazard for usual industrial or commercial handling.

Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .

NFPA

Health:0
Flammability:0
Instability:0

SDS Documents

Documents

Promotions

Promotions