missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, HPLC Grade, Ricca Chemical

ACS Reagent Grade, Suitable for Liquid Chromatography

Supplier:  Ricca Chemical Company 915332

 View more versions of this product

Catalog No. 9153-32

Explore available promotions

Description

Description

Specifications

Specifications

Water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
100%
Amber Glass Bottle
7732-18-5
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
ACS/HPLC
1 L

Chemical Identifiers

7732-18-5
18.015
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions