missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, HPLC Grade, Ricca Chemical

ACS Reagent Grade, Suitable for Liquid Chromatography

$110.25 - $232.41

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
9153-32
View Documents
Ricca Chemical Company
915332
1 L Amber Glass Bottle
Each for $110.25
 
9153-1
SDS Accessories View Documents
Ricca Chemical Company
91531
4 L Amber Glass Bottle
Each for $232.41
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.015
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane

Specifications

7732-18-5
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
ACS/HPLC
1 L
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
100%
Amber Glass Bottle
Water
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'