National Safety Apparel™ Water Resistant Cryogenic Gloves - Elbow Length
National Safety Apparel™ Water Resistant Cryogenic Gloves provide protection from ultra-cold environments in research labs, frozen food processing, dry ice blasting, and more.
Supplier:  National Safety Apparel™ G99CRBERLGEL
 View more versions of this product
Catalog No. 17-355-055
Explore available promotions
Description
Description
  • Gloves offer protection from ultra-cold environments in a wide variety of applications
  • Teflon™ Laminated Nylon with 3M™ Scotchlite™ Thinsulate™ liner
  • Water resistant, breathable membrane for additional splash protection
  • Made in the USA
Specifications
Specifications
Elbow Length
Specific
Cryogenic Gloves
Ultra-Cold Environments such as Research Labs, Frozen Food Processing, and Dry Ice Blasting
3M™ Thinsulate™
18 in.
Blue
Nylon
Large
Knit
G99CRBER_ _EL
45.7 cm