Watson-Marlow Manifold Tubing - Marprene
Double pump segment manifold tubing — use one, transfer to the other for double life.
Supplier:  Watson-Marlow 979014200
 View more versions of this product
Catalog No. 14-283-177
Explore available promotions
Description
Description
  • Made from autoclavable marprene
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
Specifications
Specifications
1/20 in.
Marprene
0.8 mm
15m/Pk.
Yellow
1.42 mm
0.03 in.
323S/MC peristaltic pump
0.14 to 9.9 (four roller pump), 0.13 to 8.8 (eight roller pump)mL/min.
Double Segment Tubing