Watson-Marlow Manifold Tubing - Marprene
Double pump segment manifold tubing — use one, transfer to the other for double life.
Supplier: Watson-Marlow 9790185000
Description
- Made from autoclavable marprene
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Specifications
7/100 in. | |
Marprene | |
0.8 mm | |
6 Pk | |
Green/Green |
1.85 mm | |
0.03 in. | |
323S/MC peristaltic pump | |
0.31 to 22 (four roller pump), 0.24 to 17 (eight roller pump)mL/min. | |
Double Segment Tubing |