Watson-Marlow Manifold Tubing - Marprene
Double pump segment manifold tubing — use one, transfer to the other for double life.
Supplier: Watson-Marlow 9790238000
Description
- Made from autoclavable marprene
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Specifications
9/100 in. | |
Marprene | |
0.8 mm | |
6 Pk. | |
Purple/Black |
2.29 mm | |
0.03 in. | |
323S/MC peristaltic pump | |
0.47 to 33 (four roller pump), 0.34 to 24 (eight roller pump)mL/min. | |
Double Segment Tubing |