missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BlueMed WAVE Shoe Covers Without Non-Skid Print
Disposable non-skid shoe covers are lightweight, breathable and provide splash protection. Ideal for transitioning between clean and unclean areas. Choose from a variety of sizes and packaging options.
$165.03 - $165.03
Specifications
Material | Polypropylene |
---|---|
Latex Free | Yes |
Color | Medical Blue |
Disposable | Single-use |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Size | Packaging Type | Packaging Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Packaging Type | Packaging Quantity | Price | Quantity | |||
18009152
|
BlueMed
SHS0102BF |
Universal | 100/Bag, 10 Bags/Cs. | 1000/Cs. |
N/A
|
|
|||
Description
- Made of a polypropylene fabric
- Shoe covers are 100% latex and silicone free
- Ideal use in medical and pharmaceutical environments, GMP facilities, pre-cleanroom gowning zones and general hygiene
- High quality fabric and strong welding construction
- Triple-elastic runs that allow for a smooth wrap around the foot to prevent tripping
- Available in 2 sizes: Universal to fit most shoe sizes, XL for bulkier or steel toe boots
- Available in several packaging options bags or dispensing boxes
- Non-Sterile, No anti-skid pattern on sole
Specifications
Polypropylene | |
Medical Blue | |
Non-sterile | |
Plain | |
Ultrasonically Welded | |
35 gsm | |
Controlled Environments Shoe Covers |
Yes | |
Single-use | |
Elastic Ankle | |
Triple-elastic | |
Medical, Pharmaceutical, GMP Facilities, Cleanroom, General PPE | |
Splash |