Wearwell™ Diamond-Plate SpongeCote™ Mat
A favorite among standing workers
Supplier:  Wearwell™ 416.1516X2X3BK
 View more versions of this product
Catalog No. 19-130-3545
Explore available promotions
Description
Description
  • 0.38 in. (0.95cm) thick, super resilient, Nitricell™ sponge base helps relieve fatigue
  • Nitricell is a special nitrile rubber/PVC compound designed for extreme comfort, immediate recovery and long life
  • Diamond embossed, 100% vinyl surface provides extra traction; self extinguishing, chemical and abrasion resistant
  • Beveled safety edges
  • Recycled: Item contains recycled content as described in the item detail
Order Info
Additional colored borders available. Contact your Fisher Scientific Customer Service Representative for details.
Specifications
Specifications
91.4 x 61 cm (3 x 2 ft.)
Black
Nitrile Rubber Composite Sponge
0.937 in.