Wearwell™ Diamond-Plate SpongeCote™ Mat
A favorite among standing workers
$105.87 - $599.01
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Thickness (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Thickness (English) | Price | Quantity | |||
17-986-124
|
Wearwell™
415.916X2X3BK |
Black | 91.4 x 61 cm (3 x 2 ft.) | 0.56 in. |
Each for $116.11
|
|
|||
17-986-125
|
Wearwell™
415.916X2X3BYL |
Black/Yellow | 91.4 x 61 cm (3 x 2 ft.) | 0.56 in. |
Each for $121.88
|
|
|||
17-986-126
|
Wearwell™
415.916X3X5BK |
Black | 152.4 x 91.4 cm (5 x 3 ft.) | 0.56 in. |
Each for $235.04
|
|
|||
17-986-127
|
Wearwell™
415.916X3X5BYL |
Black/Yellow | 152.4 x 91.4 cm (5 x 3 ft.) | 0.56 in. |
Each for $268.02
|
|
|||
19-130-3545
|
Wearwell™
416.1516X2X3BK |
Black | 91.4 x 61 cm (3 x 2 ft.) | 0.937 in. |
Each for $252.44
|
|
|||
19-150-0084
|
Wearwell™
415.916X3X10BYL |
Black/Yellow | 254 x 91 cm (10 x 3 ft.) | 0.56 in. |
Each for $599.01
|
|
|||
19-150-0083
|
Wearwell™
415.916X3X10BK |
Black | 254 x 91 cm (10 x 3 ft.) | 0.56 in. |
Each for $537.88
|
|
|||
19-150-0093
|
Wearwell™
415.916X3X10GY |
Gray | 254 x 91 cm (10 x 3 ft.) | 0.56 in. |
Each for $580.09
|
|
|||
19-150-0094
|
Wearwell™
415.916X2X3GY |
Gray | 91.4 x 61 cm (3 x 2 ft.) | 0.56 in. |
Each for $105.87
|
|
|||
19-150-0095
|
Wearwell™
415.916X3X5GY |
Gray | 152.4 x 91.4 cm (5 x 3 ft.) | 0.56 in. |
Each for $261.26
|
|
|||
Description
- 0.38 in. (0.95cm) thick, super resilient, Nitricell™ sponge base helps relieve fatigue
- Nitricell is a special nitrile rubber/PVC compound designed for extreme comfort, immediate recovery and long life
- Diamond embossed, 100% vinyl surface provides extra traction; self extinguishing, chemical and abrasion resistant
- Beveled safety edges
- Recycled: Item contains recycled content as described in the item detail
Order Info
Additional colored borders available. Contact your Fisher Scientific Customer Service Representative for details.
Specifications
Nitrile Rubber Composite Sponge |