Welch™ DirecTorr™ Gold Synthetic Pump Oil
Designed for excellent resistance when pumping corrosive gases or vapors
Supplier:  Welch™ 8995G15
 View more versions of this product
Catalog No. 01-184-105
Explore available promotions
Description
Description
Designed for excellent resistance when pumping corrosive gases or vapors
Enables excellent vacuum performance in both belt- and direct-driven vacuum pumps under harsh condition. Synthetic base stock has no aromatic compounds or sulfur. This purity inhibits varnishing, sludging and carbon buildup when pumping corrosives.
  • Double-distilled synthetic hydrocarbon oil
  • High stability in chemical environments with exceptionally low vapor pressure
  • Miscible with conventional hydrocarbon oils and can be used without rebuilding the pump
  • No additives or inhibitors
Specifications
Specifications
Gold Vacuum Pump Oil
Recommended for ChemStar pump, belt-driven or direct-drive vacuum pumps
1 gal.
Oil enables long service life and superior vacuum pump performance in applications where corrosive gases and vapors may be ingested
1 Ea.
3.78 L
Product Suggestions
Product Suggestions