missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wells Lamont™ Jomac™ Kelklave Autoclave Gloves

Jomac™ Kelklave Autoclave Gloves featuring loop-out terrycloth construction are comfortable and flexible, while also providing protection from temperatures up to 250°F.

Supplier:  Wells Lamont™ 42211

 View more versions of this product

Catalog No. 19-013586



Explore available promotions

Description

Description

Made from high-loop terry cloth, a natural insulator

  • Loop-out terry cloth is flexible and comfortable
  • Gauntlet cuff extends burn protection up the arm
  • Heat resistance up to 250°F
  • Ideal for autoclaves and metal handling
Specifications

Specifications

White
Autoclave Gloves
Clute
120°C
Gauntlet
11 in.
Terrycloth
Large
250°F
Metal Handling
422
28 cm
Videos
SDS
Documents

Documents

Promotions

Promotions