Cytiva Whatman™ Drying Rack for 903 Protein Saver Cards
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
Supplier: Cytiva 10539521
Description
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
- For use with neonatal human diagnostic testing cards
- CE Marked
- Accommodate multiple collection forms at one time
Specifications
Hook-and-Loop Mounting Sticker | |
Drying Rack | |
903 Protein Saver Card | |
10 Pack |