Cytiva Whatman™ Drying Rack for 903 Protein Saver Cards
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
$134.42 - $134.42
Specifications
Product Type Drying Rack
For Use With (Equipment) 903 Protein Saver Card
Quantity 10 Pack
View More Specs
Products 1
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
09-800-967
View Documents
Cytiva
10539521
Hook-and-Loop Mounting Sticker N/A
Description
Description
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
  • For use with neonatal human diagnostic testing cards
  • CE Marked
  • Accommodate multiple collection forms at one time
Specifications
Specifications
Drying Rack
903 Protein Saver Card
10 Pack
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'