Learn More
QIAGEN QIAcard FTA Micro
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Micro with 1 sample area per card.
Supplier:  QIAGEN WB120210
 View more versions of this product
Catalog No. 09-923-344
Only null left
Explore available promotions
Description
Description
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Micro with 1 sample area per card.
  • QIAcard FTA Micro format with 1 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
  • FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Specifications
Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Micro Cards.
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
FTA Micro Card
125 μL
Blood
1
100 Pk.
Product Suggestions
Product Suggestions