missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
QIAGEN QIAcard FTA Micro
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Micro with 1 sample area per card.
$397.32 - $692.51
Specifications
Description | Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Micro Cards. |
---|---|
Sample Type | Blood |
For Use With (Application) | Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology |
No. of Samples | 1 |
Product Type | FTA Micro Card |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
09924592
|
QIAGEN
WB120310 |
25 Pk. |
Pack of 25 Each for $397.32
|
|
||||||
09-923-344
|
QIAGEN
WB120210 |
100 Pk. |
Pack of 100 Each for $692.51
|
|
||||||
Description
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Micro with 1 sample area per card.
- QIAcard FTA Micro format with 1 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
- FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Micro Cards. | |
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology | |
FTA Micro Card |
Blood | |
1 | |
125 μL |