Cytiva Whatman™ Lens Cleaning Tissue
Provides scratch-free cleaning solutions. Whatman™ Lens Cleaning Tissue safely removes surface moisture and grease.
Supplier:  Cytiva 2105862
 View more versions of this product
Catalog No. 09-800-973
Explore available promotions
Description
Description
Provides scratch-free cleaning solutions. Whatman™ Lens Cleaning Tissue safely removes surface moisture and grease.
Soft textured strong tissue leaves no fibers. Protects lenses and other optical surfaces made from glass, quartz, or plastic from scratches.
  • Chemically pure, free of silicone and other additives
  • High absorbency leads to increased safety upon removal of surface moisture and grease
  • Thickness 0.035 to 0.040mm
  • Grade 105 sheets
Specifications
Specifications
Lens Wipe
300 x 200 mm
100 Sheets
Won't damage lenses or optical surfaces, leaves no fibers
Safe removal of surface moisture and grease.