Cytiva Whatman™ pH Indicator Papers, CF Strips
Provides an extremely clear and accurate visual pH value using four different segments of dye-impregnated indicator papers. Whatman™ pH Indicator Papers, CF Strips use dyes that are chemically bonded to the paper and cannot be leached into solution.
Supplier: Cytiva 2613991
Description
Provides an extremely clear and accurate visual pH value using four different segments of dye-impregnated indicator papers. Whatman™ pH Indicator Papers, CF Strips use dyes that are chemically bonded to the paper and cannot be leached into solution.
- Plastic supports that retain stiffness in liquids
- Paper segments on each strip have chemically bonded dyes; can't leach into solution
- Complementary indicator charts provide clear comparison
- Full-range strips (0 to 14) graduated in 1pH units; wide-range strips (4.5 to 10pH) in 0.5pH units
- Packs of 100; each strip is approximately 0.25 × 3 in. (6 × 80mm)
Specifications
0 to 14 | |
1 pH | |
100 Pack | |
80 x 6 mm | |
6 mm |
Strip | |
Plastic supports, paper segments on each strip, complementary indicator charts | |
pH Test Strip, Color Bonded | |
80 mm |