missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Funnel for 47mm Filtration Assembly

Replacement funnel for 47mm Filtration Assembly. Designed to handle up to 1000mL of liquid for analysis of particulate or microbiological contamination.

Supplier:  DWK Life Sciences 419415

 View more versions of this product

Catalog No. 14-209-131


Explore available promotions

Includes: Funnel only.

Description

Description

Designed to handle up to 1000mL of liquid for analysis of particulate or microbiological contamination

 

  • For filteration of viscous or proteinaceous solutions to produce ultra clean filtrate
  • Glass components manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements

 

Specifications

Specifications

USP type I, ASTM E 438 type I, Class A
Glass
Funnel
47mm filtration assembly
1
500 mL
Videos
SDS
Documents

Documents

Promotions

Promotions