missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask

Membrane cell culture flasks ideal for antibody and protein production

Supplier:  DWK Life Sciences WCL1000AD1

 Afficher plus de versions de ce produit

Numéro de catalogue. 02-912-745


Découvrez plus d'offres spéciales

Comprend: User manual with feeding and harvesting schedules

Description

Description

High-density cell culture flasks with dual membrane system allow for less culture maintenance and a high concentration harvest.

  • Bulk media chamber allows for longer run times without media refreshing
  • Cell compartment concentrates cells, antibodies and proteins for simplified purification
  • For suspension cell culture and adherent cell culture: Hybridoma, CHO, NS0, SF, BHK, HEK, etc.

02-912-745, 02-912-742

  • For adherent cell cultivation
  • 1000 mL media compartment
  • 15 mL cell compartment
  • Available as package of one (02-912-745) or package three (02-912-742)
02-912-746, 02-912-743
  • For suspension cell cultivation
  • 350 mL media compartment
  • 5 mL cell compartment
  • Available as package of one (02-912-746) or package five (02-912-746)
02-912-744, 02-912-741
  • For suspension cell cultivation
  • 1000 mL media compartment
  • 15 mL cell compartment
  • Available as package of one (02-912-744) or package three (02-912-741)

Spécifications

Spécifications

0.527 oz.
15 mL
Adherent Cultures
1/Cs.
35.2 oz.
1000 mL
User manual with feeding and harvesting schedules
Sterile
Videos
FDS
Documents

Documents

Offres spéciales

Offres spéciales