DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask
Membrane cell culture flasks ideal for antibody and protein production
$942.15 - $4445.53
Specifications
Includes User manual with feeding and harvesting schedules
Sterility Sterile
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Cell Capacity (Metric) Media For Use With (Application) No. per Case Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Cell Capacity (Metric) Media For Use With (Application) No. per Case Price Quantity  
02-912-741
View Documents Promotion Details
DWK Life Sciences
WCL10003
15 mL 1000 mL Suspension Cultures 3/Cs.
Pack of 3 Each for $3,373.53
02-912-744
View Documents Promotion Details
DWK Life Sciences
WCL10001
15 mL 1000 mL Suspension Cultures 1/Cs.
Pack for $1,232.12
02-912-745
View Documents Promotion Details
DWK Life Sciences
WCL1000AD1
15 mL 1000 mL Adherent Cultures 1/Cs.
Pack for $1,134.53
02-912-743
View Documents Promotion Details
DWK Life Sciences
WCL03505
5 mL 350 mL Suspension Cultures 5/Cs.
Pack of 5 Each for $4,445.53
02-912-742
View Documents Promotion Details
DWK Life Sciences
WCL1000AD3
15 mL 1000 mL Adherent Cultures 3/Cs.
Pack of 3 Each for $3,403.47
02-912-746
View Documents Promotion Details
DWK Life Sciences
WCL03501
5 mL 350 mL Suspension Cultures 1/Cs.
Pack for $942.15
Description
Description
High-density cell culture flasks with dual membrane system allow for less culture maintenance and a high concentration harvest.
  • Bulk media chamber allows for longer run times without media refreshing
  • Cell compartment concentrates cells, antibodies and proteins for simplified purification
  • For suspension cell culture and adherent cell culture: Hybridoma, CHO, NS0, SF, BHK, HEK, etc.
02-912-745, 02-912-742
  • For adherent cell cultivation
  • 1000 mL media compartment
  • 15 mL cell compartment
  • Available as package of one (02-912-745) or package three (02-912-742)
02-912-746, 02-912-743
  • For suspension cell cultivation
  • 350 mL media compartment
  • 5 mL cell compartment
  • Available as package of one (02-912-746) or package five (02-912-746)
02-912-744, 02-912-741
  • For suspension cell cultivation
  • 1000 mL media compartment
  • 15 mL cell compartment
  • Available as package of one (02-912-744) or package three (02-912-741)
Specifications
Specifications
User manual with feeding and harvesting schedules
Sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'