missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences WHEATON™ COMPLETEPAK MODPAK Amber Kit
Range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Supplier: DWK Life Sciences W991004ACPMP
Description
- Six inventory matched kits conveniently packed into a single shipper case.
- Sterilized, off-the-shelf, ready-to-use (RtU) primary packaging components
- Focuses your effort on core competencies and reduces lead time
- Streamlines process and frees up valuable clean room space USP /EP 2.9.19
- Particulate Matter in Injections, USP /EP 2.6.1,
- The Bacterial Endotoxins Test and USP / EP 2.6.14
- The Sterility Test, USP /EP 3.2.1
- Glass Containers for Pharmaceutical Use
- USP /EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
- Complete manufacturing & certificate of analysis traceability
- Ready-to-Use, Just-in-Time
- Minimum 180 day shelf life from time of shipment
- Container Closure Integrity (CCI) validation
- Available in amber for photosensitive contents
Specifications
Red | |
Borosilicate Glass | |
Single-use | |
No | |
CompletePAK ModPAK | |
Autoclavable | |
32 mm | |
USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and USP <71>/ EP 2.6.14 The Sterility Test, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations | |
13 mm |
Amber | |
Sterile | |
Compounding pharmaceuticals, Aseptic filling of injectable drug products, Drug discovery, Drug research, Freeze drying / lyophilized drug products, Large and small molecule drug packaging | |
Vial, Stopper, and Seal | |
Bromobutyl | |
Omni Flex Serum Stopper | |
278/Pk. | |
Serum Vial |