missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences WHEATON™ COMPLETEPAK MODPAK Amber Kit
Range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
$14696.38 - $35436.68
Specifications
Color | Amber |
---|---|
Material | Borosilicate Glass |
Sterility | Sterile |
Disposable | Single-use |
For Use With (Application) | Compounding pharmaceuticals, Aseptic filling of injectable drug products, Drug discovery, Drug research, Freeze drying / lyophilized drug products, Large and small molecule drug packaging |
Catalog Number | Mfr. No. | Closure Material | Diameter (Metric) Outer | Closure Color | Closure Type | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Material | Diameter (Metric) Outer | Closure Color | Closure Type | Price | Quantity | ||
15112207
|
DWK Life Sciences
W991000ACPMP |
Bromobutyl | Blue | Omni Flex Serum Stopper |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112208
|
DWK Life Sciences
W991001ACPMP |
Bromobutyl | Blue | Omni Flex Serum Stopper |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112209
|
DWK Life Sciences
W991002ACPMP |
Bromobutyl | Blue | Omni Flex Igloo Lyo Stopper |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112210
|
DWK Life Sciences
W991003ACPMP |
Bromobutyl | Red | Omni Flex Serum Stopper |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112211
|
DWK Life Sciences
W991004ACPMP |
Bromobutyl | Red | Omni Flex Serum Stopper |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112212
|
DWK Life Sciences
W991005ACPMP |
Bromobutyl | Red | Omni Flex Igloo Lyo Stopper |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112213
|
DWK Life Sciences
W991006ACPMP |
Bromobutyl | 22 mm | Blue | Omni Flex Serum Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112214
|
DWK Life Sciences
W991007ACPMP |
Bromobutyl | 22 mm | Blue | Omni Flex Serum Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112215
|
DWK Life Sciences
W991008ACPMP |
Bromobutyl | 22 mm | Blue | Omni Flex Igloo Lyo Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112216
|
DWK Life Sciences
W991009ACPMP |
Bromobutyl | 22 mm | Red | Omni Flex Serum Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112217
|
DWK Life Sciences
W991010ACPMP |
Bromobutyl | 22 mm | Red | Omni Flex Serum Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112218
|
DWK Life Sciences
W991011ACPMP |
Bromobutyl | 22 mm | Red | Omni Flex Igloo Lyo Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112219
|
DWK Life Sciences
W991012ACPMP |
Bromobutyl | 24 mm | Blue | Omni Flex Serum Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112220
|
DWK Life Sciences
W991013ACPMP |
Bromobutyl | 24 mm | Blue | Omni Flex Serum Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112221
|
DWK Life Sciences
W991014ACPMP |
Bromobutyl | 24 mm | Blue | Omni Flex Igloo Lyo Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112222
|
DWK Life Sciences
W991015ACPMP |
Bromobutyl | 24 mm | Red | Omni Flex Serum Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112223
|
DWK Life Sciences
W991016ACPMP |
Bromobutyl | 24 mm | Red | Omni Flex Serum Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15112224
|
DWK Life Sciences
W991017ACPMP |
Bromobutyl | 24 mm | Red | Omni Flex Igloo Lyo Stopper |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Six inventory matched kits conveniently packed into a single shipper case.
- Sterilized, off-the-shelf, ready-to-use (RtU) primary packaging components
- Focuses your effort on core competencies and reduces lead time
- Streamlines process and frees up valuable clean room space USP /EP 2.9.19
- Particulate Matter in Injections, USP /EP 2.6.1,
- The Bacterial Endotoxins Test and USP / EP 2.6.14
- The Sterility Test, USP /EP 3.2.1
- Glass Containers for Pharmaceutical Use
- USP /EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
- Complete manufacturing & certificate of analysis traceability
- Ready-to-Use, Just-in-Time
- Minimum 180 day shelf life from time of shipment
- Container Closure Integrity (CCI) validation
- Available in amber for photosensitive contents
Specifications
Amber | |
Sterile | |
Compounding pharmaceuticals, Aseptic filling of injectable drug products, Drug discovery, Drug research, Freeze drying / lyophilized drug products, Large and small molecule drug packaging | |
Vial, Stopper, and Seal | |
Bromobutyl | |
USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and USP <71>/ EP 2.6.14 The Sterility Test, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations |
Borosilicate Glass | |
Single-use | |
No | |
CompletePAK ModPAK | |
Autoclavable | |
Serum Vial |