DWK Life Sciences Wheaton™ Dounce Tissue Grinders
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
Supplier:  DWK Life Sciences 357544
 View more versions of this product
Catalog No. 06-435B
Explore available promotions
Description
Description
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
  • Sample covers the pestle ball, preventing heat buildup by reducing friction against the mortar
  • Mortar features large reservoir and pouring lip
  • Supplied with “loose” and “tight” pestles
  • Components are made of low extractable borosilicate glass
Specifications
Specifications
0.5 oz.
Borosilicate Glass
Designed to retain a high percentage of cell nuclei, mitochondria and Proteins in soft tissues or cell suspensions
210mm
94mm
22mm
15 mL
Dounce Tissue Grinder
8.26 in.
3.7 in.
0.86 in.
0.5 oz., 15mL