missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Dounce Tissue Grinders
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
$369.38 - $675.06
Specifications
Material | Borosilicate Glass |
---|---|
Product Type | Dounce Tissue Grinder |
For Use With (Application) | Designed to retain a high percentage of cell nuclei, mitochondria and Proteins in soft tissues or cell suspensions |
Catalog Number | Mfr. No. | Capacity | Length (English) | Length (English) Tube | Tube Diameter (English) Outside | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Length (English) | Length (English) Tube | Tube Diameter (English) Outside | Price | Quantity | ||
06-435B
|
DWK Life Sciences
357544 |
0.5 oz., 15mL | 8.26 in. | 3.7 in. | 0.86 in. |
Each for $458.97
Case of 2 Each for $917.94
|
|
||
06-435A
|
DWK Life Sciences
357542 |
0.23 oz., 7mL | 6.8 in. | 3.22 in. | 0.51 in. |
Each for $369.38
Case of 2 Each for $738.76
|
|
||
06-434
|
DWK Life Sciences
357538 |
0.03 oz., 1mL | 4.92 in. | 1.8 in. | 0.43 in. |
Case of 2 Each for $675.06
|
|
||
06-435C
|
DWK Life Sciences
357546 |
1.35 oz., 40mL | 11.2 in. | 5.5 in. | 1.25 in. |
Each for $517.00
Case of 2 Each for $1,034.00
|
|
||
Description
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
- Sample covers the pestle ball, preventing heat buildup by reducing friction against the mortar
- Mortar features large reservoir and pouring lip
- Supplied with “loose” and “tight” pestles
- Components are made of low extractable borosilicate glass
Order Info
- Tight pestle clearance: 0.0020 ±0.001 in. (0.05 ±0.025mm)
- Loose pestle clearance: 0.114 ±0.001 in. (0.114 ±0.025mm)
Specifications
Borosilicate Glass | |
Dounce Tissue Grinder | |
Designed to retain a high percentage of cell nuclei, mitochondria and Proteins in soft tissues or cell suspensions |