DWK Life Sciences Wheaton™ Glass Sample Vials in Vial File™
Vial File provides a compact, easily accessible means of sample storage
Supplier:  DWK Life Sciences W224696
 View more versions of this product
Catalog No. 02-912-512
Explore available promotions
Description
Description
Vial File provides a compact, easily accessible means of sample storage
Caps attached to vials.
  • Polystyrene case with partitions, foam inserts and alphanumeric indexing card helps organize and protect vials
  • Choice of solid- or open-top black phenolic caps: open caps facilitate sample retrieval with syringe
  • Vials and caps autoclavable
Clear or Amber Vials
  • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
  • Amber vials made from borosilicate glass that protects light-sensitive products
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements of light transmission.
Specifications
Specifications
PTFE/14B Rubber
15 mm
4 mL
No
1.08 dr.
Autoclavable
48 mm
Amber
Borosilicate Glass
Used for storing and transporting vials
Screw
Phenolic
Screw Cap
USP Type I