DWK Life Sciences Wheaton™ Glass Sample Vials in Vial File™
Vial File provides a compact, easily accessible means of sample storage
Supplier:  DWK Life Sciences 224891
 View more versions of this product
Catalog No. 03-336
Explore available promotions
Description
Description
Vial File provides a compact, easily accessible means of sample storage
Caps attached to vials.
  • Polystyrene case with partitions, foam inserts and alphanumeric indexing card helps organize and protect vials
  • Choice of solid- or open-top black phenolic caps: open caps facilitate sample retrieval with syringe
  • Vials and caps autoclavable
Clear or Amber Vials
  • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
  • Amber vials made from borosilicate glass that protects light-sensitive products
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements of light transmission.
Specifications
Specifications
14B Rubber
12 mm
2.0 mL
No
0.5 dr.
Autoclavable
38 mm
Clear
Borosilicate Glass (Low Extractable)
Used for storing and transporting vials
Screw
Phenolic
Screw Cap
ASTM E438, USP Type I, Type I, Class A