DWK Life Sciences Wheaton™ I-LOC Closures
Eliminate use of separate aluminum seal and rubber stopper
Supplier:  DWK Life Sciences 24067601
 View more versions of this product
Catalog No. 06-440-48
Explore available promotions
Description
Description
Eliminate use of separate aluminum seal and rubber stopper
  • Polypropylene closures
  • Gray bromobutyl stoppers
Specifications
Specifications
Bromobutyl
Polypropylene
Screw Cap
20-400 Screw Thread Vials
Black
Autoclavable
20 mm