DWK Life Sciences Wheaton™ I-LOC Closures
Eliminate use of separate aluminum seal and rubber stopper
$111.50 - $122.33
Specifications
Material (Liner) Bromobutyl
Material Polypropylene
Autoclavable Autoclavable
Type Screw Cap
View More Specs
Products 3
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
06-440-48
View Documents Promotion Details
DWK Life Sciences
24067601
Black
Case of 100 Each for $111.50
06-440-50
View Documents Promotion Details
DWK Life Sciences
24067603
Red
Case of 100 Each for $122.33
06-440-49
View Documents Promotion Details
DWK Life Sciences
24067602
White, Gray
Case of 100 Each for $115.29
Description
Description
Eliminate use of separate aluminum seal and rubber stopper
  • Polypropylene closures
  • Gray bromobutyl stoppers
Specifications
Specifications
Bromobutyl
Polypropylene
Autoclavable
Screw Cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'