DWK Life Sciences Wheaton™ Magna-Flex™ Spinner Flasks
Produce high-yield cultures by maintaining even circulation while keeping microcarriers in suspension
Supplier:  DWK Life Sciences 356830
 View more versions of this product
Catalog No. 06-416-1A
Explore available promotions
Description
Description
  • Borosilicate glass flasks
  • Two large side arms with screw-cap closures
  • Flex-type, bulb-shaped glass impeller rotates from a fixed position above the liquid level around an indentation in the base of the flask
  • Graduated
  • Removable stainless-steel pin immobilizes the impeller during handling or decanting to prevent damage to cells or microcarriers
  • Autoclavable flasks and impellers for selected sizes can also be ordered separately
  • U.S. Patent No. 3,854,704
Specifications
Specifications
125 mL
Flask with impeller
Autoclavable
51-440
Borosilicate Glass
33-430
Yes