DWK Life Sciences Wheaton™ Magna-Flex™ Spinner Flasks
Produce high-yield cultures by maintaining even circulation while keeping microcarriers in suspension
$496.00 - $2161.68
Catalog Number | Mfr. No. | Capacity (Metric) | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Includes | Price | Quantity | ||||
12141551
|
DWK Life Sciences
356834 |
1000 mL | Flask with impeller |
Each for $1,687.09
|
|
||||
06-416-1C
|
DWK Life Sciences
356832 |
500 mL | Flask with impeller |
Case for $1,178.09
|
|
||||
06-416-1A
|
DWK Life Sciences
356830 |
125 mL | Flask with impeller |
Case for $791.15
|
|
||||
06-416-1B
|
DWK Life Sciences
356831 |
250 mL | Flask with impeller |
Case for $882.74
|
|
||||
06-416-7C
|
DWK Life Sciences
356916 |
125 mL | Flask only |
Each for $496.00
|
|
||||
12141552
|
DWK Life Sciences
356837 |
3000 mL | Flask with impeller |
Each for $2,161.68
|
|
||||
Description
- Borosilicate glass flasks
- Two large side arms with screw-cap closures
- Flex-type, bulb-shaped glass impeller rotates from a fixed position above the liquid level around an indentation in the base of the flask
- Graduated
- Removable stainless-steel pin immobilizes the impeller during handling or decanting to prevent damage to cells or microcarriers
- Autoclavable flasks and impellers for selected sizes can also be ordered separately
- U.S. Patent No. 3,854,704
Specifications
Yes |