DWK Life Sciences Wheaton™ Prescored Gold-Band™ Clear Ampules
Prescored to eliminate need for filing
Supplier:  DWK Life Sciences 176780
 View more versions of this product
Catalog No. 01-215D
Explore available promotions
Description
Description
Prescored to eliminate need for filing
  • Low-extractable borosilicate glass
  • Gold-Band feature indicates prescoring
  • Stems can be pull sealed or tip sealed
Specifications
Specifications
10 mL
Prescored Ampule
1.9 cm
10.7 cm
USP Type 1, ASTM Type I, Class A
Borosilicate Glass
Clear
0.74 in.
4.21 in.
Videos
SDS
Documents
Documents
Accessories