missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Snap-on Style Stoppers for Vials and Serum Bottles
For maximum sealing of vials with blow-back glass finish
Supplier: DWK Life Sciences W224100342
Description
For maximum sealing of vials with blow-back glass finish
- Made of gray bromobutyl
Not recommended for vials with a straight-walled finish.
Specifications
Gray | |
3cm | |
Fits 1.5cm I.D., 3cm O.D. mouth of vials and serum bottles | |
Autoclavable |
Plug Stopper | |
Bromobutyl Rubber | |
50 Durometer |