DWK Life Sciences Wheaton™ Snap-on Style Stoppers for Vials and Serum Bottles
For maximum sealing of vials with blow-back glass finish
$484.65 - $1,712.41
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Material | Hardness | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Material | Hardness | Price | Quantity | |||
03-084-141
|
DWK Life Sciences
W224100342 |
3cm | Bromobutyl Rubber | 50 Durometer |
N/A
|
|
|||
02-985-135
|
DWK Life Sciences
W224100081 |
1.3cm | Chlorobutyl Rubber | 45 Durometer |
N/A
|
|
|||
Description
For maximum sealing of vials with blow-back glass finish
- Made of gray bromobutyl
Not recommended for vials with a straight-walled finish.
Specifications
Autoclavable | |
Gray | |
Plug Stopper |