Socorex Acura™ Manual Pipets 826 XS TwiXS pack from DWK Life Sciences™
Two adjustable volume pipettes available in six different packs
Supplier:  Socorex W810307TXS
 View more versions of this product
Catalog No. 13-709-410
Explore available promotions
Description
Description
  • Two adjustable volume pipettes
  • Free shelf pipette holder included
  • Volume combinations up to 1000μL
  • Six different packs available
Specifications
Specifications
CE, IVD 98/79 EEC
Acura™ 826 XS
Pipette
Ergonomic
Yes
Autoclavable
Socorex™ Acura™
0.5 to 10, 10 to 100 μL
Shelf pipette holder, pipette tip samples, Silicone Grease, Q.C. certificates and operating instructions
3 Years