DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Sleeve Stopper
Provide airtight covers
Supplier:  DWK Life Sciences 224100320
 View more versions of this product
Catalog No. 06-451-60
Explore available promotions
Description
Description
  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
Specifications
Red
3 cm
Rubber
40 Durometer
Sleeve Stopper
Multi-puncture applications
Fits 1.5cm I.D., 3cm O.D. mouth of serum bottles and vials
Autoclavable