missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Sleeve Stopper

Provide airtight covers

$587.06 - $2203.38

Specifications

Autoclavable Autoclavable
Color Red
Material Rubber
Product Type Sleeve Stopper
For Use With (Application) Multi-puncture applications
View More Specs

Products 4
Catalog Number Mfr. No. Color Material For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Color Material For Use With (Equipment) Price Quantity  
06-406-10A
View Documents Promotion Details
DWK Life Sciences
224100060
Red Rubber Fits 0.7cm I.D., 1.3cm O.D. mouth of serum bottles and vials
Case of 1000 Each for $1,444.50
 
06-447A
View Documents Promotion Details
DWK Life Sciences
224100020
Red Rubber Fits 0.5cm I.D., 1.1cm O.D. mouth of serum bottles and vials
Case of 1000 Each for $587.06
 
06-451-60
View Documents Promotion Details
DWK Life Sciences
224100320
Red Rubber Fits 1.5cm I.D., 3cm O.D. mouth of serum bottles and vials
Case of 1000 Each for $2,203.38
 
06-406-10B
View Documents Promotion Details
DWK Life Sciences
224100161
Red Rubber Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials
Case of 1000 Each for $1,295.15
 
Description

Description

  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications

Specifications

Autoclavable
Rubber
Multi-puncture applications
Red
Sleeve Stopper
40 Durometer
SDS
missing translation for 'documents'

missing translation for 'documents'