Corepoint Scientific™ White Diamond Series Laboratory and Medical Stainless Steel Freezer
This high-performance White Diamond Series Laboratory and Medical Stainless Steel Freezer provides quality cold storage in laboratory settings. Solid, stainless steel construction adds to the professionalism and long-term durability of this unit.
$16758.65 - $23560.83
Specifications
Type Upright Freezer
Temperature Sensor Microprocessor Temperature Controller
Temperature Stability ±0.1°C
Door Style Solid Swing
Interior Aluminum, Stainless Steel
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Temperature Range No. of Doors Price Quantity  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Temperature Range No. of Doors Price Quantity  
22651333
View Documents
Corepoint Scientific™
NSWDF231SSS0A
23 cu. ft. 651 L -15°C to -25°C 1 N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-651-362
View Documents
Corepoint Scientific™
NSWDF231SSS0A3
23 cu. ft. 651 L -27°C to -35°C 1 N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22651334
View Documents
Corepoint Scientific™
NSWDF492SSS0A
49 cu. ft. 1388 L -15°C to -25°C 2 N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Microprocessor temperature controller
  • Touch screen digital display
  • Integrated temperature and product safety alarms
  • Password-protected control panel
  • USB port for data transfer and software updates
  • Digital 24-hour temperature chart
  • Temperature data logging
  • Probe access port
  • Keyed door lock
  • Adjustable shelving and interior lighting
  • Natural hydrocarbon refrigerant
Specifications
Specifications
Upright Freezer
±0.1°C
Aluminum, Stainless Steel
Upright
Yes
Fan-forced
Probe Access Port
Automatic
Alarm Module, Battery Back-up
115 V
Natural Hydrocarbon Refrigerant
Microprocessor Temperature Controller
Solid Swing
ETL
Touch Screen Digital Temperature Display
Silver
Cold Storage
3 year parts and labor warranty, plus an additional 2 year compressor parts warranty
87 in.
NEMA 5-15
H-drive
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'