Fisherbrand™ White Polypropylene Caps
Fisherbrand™ White Polypropylene Caps include a clear writing surface for easy identification.
 View more versions of this product
Catalog No. FB02912291A
Explore available promotions
Description
Description
  • Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
  • Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Specifications
White
White
Pulp/Vinyl
48-400 mm
576
Lined Cap
Polypropylene