Fisherbrand™ White Polypropylene Caps
Fisherbrand™ White Polypropylene Caps include a clear writing surface for easy identification.
$173.32 - $643.86
Catalog Number | Mfr. No. | Closure Size | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Size | Quantity | Price | Quantity | ||||
FB02912064A
|
N/A | 13-425 mm | 500 |
Case of 500 Each for $349.71
|
|
||||
FB02912291A
|
N/A | 48-400 mm | 576 |
Case of 576 Each for $643.86
|
|
||||
FB02912332A
|
N/A | 48-400 mm | 1700 |
Case of 1700 Each for $419.64
|
|
||||
FB02912088A
|
N/A | 70-400 mm | 144 |
Case of 144 Each for $173.32
|
|
||||
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
White | |
Polypropylene |