missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Wide Mouth Glass Reagent Bottle with Glass Stopper
Made from high quality borosilicate 3.3 glass. Extremely durable with a low temperature gradient and superior chemical resistance.
$20.24 - $134.71
Specifications
Color | Clear |
---|---|
Description | Wide neck |
For Use With (Application) | Holding liquids |
Includes | Interchangeable, tight fitting, hollow glass stopper (socket size 24/29) |
Material | Borosilicate Glass |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | |||||
S40893
|
Eisco™
CH0163A |
60 mL | 24/29 mm |
Each for $20.24
|
|
|||||
S40894
|
Eisco™
CH0163B |
125 mL | 29/32 mm |
Each for $25.76
|
|
|||||
S40895
|
Eisco™
CH0163C |
250 mL | 34/35 mm |
Pack for $32.56
|
|
|||||
S40896
|
Eisco™
CH0163D |
500 mL | 45/40 mm |
Pack for $67.41
|
|
|||||
S40899
|
Eisco™
CH0163E |
1000 mL | 50/42 mm |
Pack for $116.35
|
|
|||||
S40901
|
Eisco™
CH0163F |
2000 mL | 55/44 mm |
Pack for $134.71
|
|
|||||
Description
Warranty
1 year limited manufacturer warranty
Specifications
Clear | |
Holding liquids | |
Borosilicate Glass | |
Clear | |
Autoclavable | |
No | |
Cylindrical | |
Liquids | |
Glass |
Wide neck | |
Interchangeable, tight fitting, hollow glass stopper (socket size 24/29) | |
Non-sterile | |
Yes | |
Reusable | |
Narrow | |
Reagent Bottle | |
Unlabeled | |
Stopper |