missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Wide Mouth Glass Reagent Bottle with Glass Stopper

Made from high quality borosilicate 3.3 glass. Extremely durable with a low temperature gradient and superior chemical resistance.

Supplier:  Eisco™ CH0163F

 View more versions of this product

Catalog No. S40901

Explore available promotions

Description

Description

  • Clear reagent bottle made from heavy duty borosilicate 3.3 glass
  • Wide mouth with interchangeable hollow borosilicate glass stopper
  • No graduations
  • Autoclavable

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

2000 mL
67.6 oz.
Wide neck
Interchangeable, tight fitting, hollow glass stopper (socket size 24/29)
18.8 cm
Non-sterile
18.8 cm
Yes
Reusable
Narrow
38.4 cm
For kitchens, crafting, and for storage
Unlabeled
55/44 mm
Stopper
2000 mL
Clear
Holding liquids
Borosilicate Glass
1
1.17 kg
Clear
Autoclavable
No
Cylindrical
Reagent Bottle
15.11 in.
Glass
18.8 cm
SDS
Documents

Documents

Promotions

Promotions