Poltex Wipe Canister and Glove Box Holder
A break-resistant PETG wipe canister and glove box holder that holds two boxes of gloves and a wipe tub.
Supplier: Poltex GBSTUFW
Description
- Rubber bumpons on base for non-slip placement on countertop.
- Choose from wall mount/countertop placement (screws/anchors included), magnet mount (powerful magnets included), or tape mount (VHB tape included).
Specifications
Wipe Dispenser | |
PETG | |
Wall Mount | |
2 Boxes |