Fisherbrand™ Flask Shaker, Single Speed
High performance flask shaker with expansion capabilities to meet all of your laboratory needs.
 View more versions of this product
Catalog No. 14400600
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Single Speed with Timer
  • Accommodates 8 Vessels
  • Shafts and Clamps Included
  • 385 OSC per Minute
  • Max tested Load at 10 LBS
Specifications
Specifications
0 to 15°
115 V
10 lb.
70 lb.
8
385 rpm
8 x 250 mL vessels
60 Hz
Side Arm Shafts, Clamps, Power Cord, Manual, Allen Wrench, Lever Arm Break
0 to 60 min.